missing translation for 'onlineSavingsMsg'
Learn More

6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate, 95%

Product Code. 15479025
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15479025 1 g 1g
15489025 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15479025 Supplier Thermo Scientific Alfa Aesar Supplier No. H35567.03
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 138642-97-4 | C8H5ClN2O2 | 196.59 g/mol

6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138642-97-4
Molecular Formula C8H5ClN2O2
Molecular Weight (g/mol) 196.59
MDL Number MFCD07021509
InChI Key LXRDUEDQFSBROM-UHFFFAOYSA-N
Synonym 6-chloroimidazo 1,2-a pyridine-3-carboxylic acid, imidazo 1,2-a pyridine-3-carboxylic acid, 6-chloro
PubChem CID 16413639
IUPAC Name 6-chloroimidazo[1,2-a]pyridine-3-carboxylic acid
SMILES OC(=O)C1=CN=C2C=CC(Cl)=CN12

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 196.59 (Anhydrous)
Percent Purity 95%
Chemical Name or Material 6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.