missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2-fluoro-3-methoxybenzamide, 97%, Thermo Scientific™

Product Code. p-7030785
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15404247

Brand: Thermo Scientific Alfa Aesar H31638.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886761-75-7
Molecular Formula C8H7ClFNO2
Molecular Weight (g/mol) 203.597
MDL Number MFCD06660354
InChI Key AZUOJQIOSGRMMS-UHFFFAOYSA-N
Synonym 6-chloro-2-fluoro-3-methoxybenzenecarboxamide
PubChem CID 17999360
IUPAC Name 6-chloro-2-fluoro-3-methoxybenzamide
SMILES COC1=C(C(=C(C=C1)Cl)C(=O)N)F

Specifications

Melting Point 143°C to 145°C
Quantity 1 g
Formula Weight 203.6
Percent Purity 97%
Chemical Name or Material 6-Chloro-2-fluoro-3-methoxybenzamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.