Learn More
6-Chloro-2,3-difluorobenzyl alcohol, 97%, Thermo Scientific™
Product Code. 15436947
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15436947 5 g 5g
15426947 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15436947 Supplier Thermo Scientific Alfa Aesar Supplier No. H34041.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 887585-70-8
Molecular Formula C7H5ClF2O
Molecular Weight (g/mol) 178.56
MDL Number MFCD07368108
InChI Key QOCZAGKTHBSPMS-UHFFFAOYSA-N
Synonym 6-chloro-2,3-difluorobenzyl alcohol, 6-chloro-2,3-difluorobenzenemethanol, 6-chloro-2,3-difluorophenyl methanol
PubChem CID 29922043
IUPAC Name (6-chloro-2,3-difluorophenyl)methanol
SMILES OCC1=C(Cl)C=CC(F)=C1F
Specifications
Quantity 5 g
Formula Weight 178.57
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Chloro-2,3-difluorobenzyl alcohol