missing translation for 'onlineSavingsMsg'
Learn More

6-Bromopyrazolo[1,5-a]pyrimidine-3-carbonitrile, 98%, Thermo Scientific™

Product Code. p-7032298
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458595 1 g 1g
15468595 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15458595 Supplier Thermo Scientific Alfa Aesar Supplier No. H33701.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352637-44-6
Molecular Formula C7H3BrN4
Molecular Weight (g/mol) 223.03
MDL Number MFCD00461392
InChI Key IDYCEZBYNPADDN-UHFFFAOYSA-N
Synonym 6-bromopyrazolo 1,5-a pyrimidine-3-carbonitrile, 6-bromo-pyrazolo 1,5-a pyrimidine-3-carbonitrile, 6-bromo-3-cyano-pyrazolo 1,5-a pyrimidine, pyrazolo 1,5-a pyrimidine-3-carbonitrile,6-bromo, 6-bromo-3-cyanopyrazolo 1,5-a pyrimidine, pyrazolo 1,5-a pyrimidine-3-carbonitrile, 6-bromo, 3,5-dimethoxybenzyl 4-methoxybenzyl amine hydrobromide, 6-bromo-8-hydropyrazolo 1,5-a pyrimidine-3-carbonitrile
PubChem CID 614285
IUPAC Name 6-bromopyrazolo[1,5-a]pyrimidine-3-carbonitrile
SMILES BrC1=CN2N=CC(C#N)=C2N=C1

Specifications

Melting Point 154°C to 156°C
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 223.03
Percent Purity 98%
Chemical Name or Material 6-Bromopyrazolo[1,5-a]pyrimidine-3-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.