Learn More
6-Bromo-5-methyl-3h-imidazo[4,5-b]pyridine, TRC
Specifications
Specifications
| Chemical Name or Material | 6-Bromo-5-methyl-3h-imidazo[4,5-b]pyridine |
| CAS | 28279-41-6 |
| Molecular Formula | C7H6BrN3 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C7H6BrN3/c1-4-5(8)2-6-7(11-4)10-3-9-6/h2-3H,1H3,(H,9,10,11) |
| SMILES | Cc1nc2nc[nH]c2cc1Br |
| IUPAC Name | 6-bromo-5-methyl-1H-imidazo[4,5-b]pyridine |
| Molecular Weight (g/mol) | 212.05 |
| Formula Weight | 210.9745 |