Learn More
6-Bromo-3-hydroxypyrazine-2-carboxamide, TRC
Product Code. 30138072
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138072 250 mg 250mg
30138177 500 mg 500mg
30007650 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30138072 Supplier Toronto Research Chemicals Supplier No. TRCB103355250MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 259793-88-9
Molecular Formula C5 H4 Br N3 O2
Molecular Weight (g/mol) 218.008
Synonym 2-Pyrazinecarboxamide, 6-bromo-3,4-dihydro-3-oxo- (ACI), 6-Bromo-3,4-dihydro-3-oxo-2-pyrazinecarboxamide (ACI), Pyrazinecarboxamide, 6-bromo-3,4-dihydro-3-oxo- (9CI), 6-Bromo-3-hydroxypyrazine-2-carboxamide, Br-T-705
IUPAC Name 5-bromo-2-oxo-1H-pyrazine-3-carboxamide
SMILES NC(=O)C1=NC(=CNC1=O)Br
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C5H4BrN3O2/c6-2-1-8-5(11)3(9-2)4(7)10/h1H,(H2,7,10)(H,8,11)
Formula Weight 216.949
Chemical Name or Material 6-Bromo-3-hydroxypyrazine-2-carboxamide