Learn More
6-Bromo-2-oxindole, TRC
Click to view available options
Quantity:
2 g
Unit Size:
2g
Specifications
Specifications
| Chemical Name or Material | 6-Bromo-2-oxindole |
| CAS | 99365-40-9 |
| Molecular Formula | C8H6BrNO |
| Quantity | 2 g |
| Synonym | 6-Bromo-1,3-dihydro-2H-indol-2-one, 6-Bromo-1,3-dihydroindol-2-one, 6-Bromo-2-indolinone, 6-Bromo-2-oxoindoline, 6-Bromoindol-2-one, 6-Bromooxindole |
| InChI Formula | InChI=1S/C8H6BrNO/c9-6-2-1-5-3-8(11)10-7(5)4-6/h1-2,4H,3H2,(H,10,11) |
| SMILES | Brc1ccc2CC(=O)Nc2c1 |
| IUPAC Name | 6-bromo-1,3-dihydroindol-2-one |
| Molecular Weight (g/mol) | 212.04 |
| Formula Weight | 210.9633 |