Learn More
6-Bromo-2-chloro-8-cyclopentyl-5-methylpyrido-[2,3-d]pyrimidin-7(8H)-one, TRC
Specifications
Specifications
| Chemical Name or Material | 6-Bromo-2-chloro-8-cyclopentyl-5-methylpyrido-[2,3-d]pyrimidin-7(8H)-one |
| CAS | 1016636-76-2 |
| Molecular Formula | C13H13BrClN3O |
| Quantity | 250 mg |
| Synonym | Pyrido[2,3-d]pyrimidin-7(8H)-one, 6-bromo-2-chloro-8-cyclopentyl-5-methyl- (ACI), 6-Bromo-2-chloro-8-cyclopentyl-5-methylpyrido[2,3-d]pyrimidin-7(8H)-one (ACI), 6-Bromo-2-chloro-8-cyclopentyl-5-methyl-8H-pyrido[2,3-d]pyrimidin-7-one |
| InChI Formula | InChI=1S/C13H13BrClN3O/c1-7-9-6-16-13(15)17-11(9)18(12(19)10(7)14)8-4-2-3-5-8/h6,8H,2-5H2,1H3 |
| SMILES | CC1=C(Br)C(=O)N(C2CCCC2)c3nc(Cl)ncc13 |
| IUPAC Name | 6-bromo-2-chloro-8-cyclopentyl-5-methylpyrido[2,3-d]pyrimidin-7-one |
| Molecular Weight (g/mol) | 342.62 |