Learn More
6-Amino-2,4-dihydroxypyrimidine, TRC
Product Code. 30327500
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30327500 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30327500 Supplier Toronto Research Chemicals Supplier No. TRCA6049352.5G
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 6-Amino-2,4-dihydroxypyrimidine
CAS 873-83-6
Molecular Formula C4 H5 N3 O2
Quantity 2.5 g
Synonym 2,4(1H,3H)-Pyrimidinedione, 6-amino-, Uracil, 6-amino- (6CI,7CI,8CI), 6-Amino-2,4(1H,3H)-pyrimidinedione, 2,4-Dihydroxy-6-aminopyrimidine, 4-Amino-2,6-dihydroxypyrimidine, 4-Aminouracil, 6-Amino-2,4-dihydroxypyrimidine, 6-Amino-2,4-dioxo-1,2,3,4-tetrahydropyrimidine, 6-Aminopyrimidine-2,4(1H,3H)-dione, 6-Aminopyrimidine-2,4-diol, 6-Aminouracil, NSC 15919, NSC 7367
InChI Formula InChI=1S/C4H5N3O2/c5-2-1-3(8)7-4(9)6-2/h1H,(H4,5,6,7,8,9)
SMILES Nc1cc(O)nc(O)n1
IUPAC Name 6-aminopyrimidine-2,4-diol
Molecular Weight (g/mol) 127.1
Formula Weight 127.0382