Learn More
6-[[2-(tert-Butyldimethylsilyloxy)ethyl]methylamino]pyridin-3-ol, TRC
Product Code. 30336184
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336184 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30336184 Supplier Toronto Research Chemicals Supplier No. TRCB691445500MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 6-[[2-(tert-Butyldimethylsilyloxy)ethyl]methylamino]pyridin-
CAS 326496-02-0
Molecular Formula C14 H26 N2 O2 Si
Quantity 500 mg
InChI Formula InChI=1S/C14H26N2O2Si/c1-14(2,3)19(5,6)18-10-9-16(4)13-8-7-12(17)11-15-13/h7-8,11,17H,9-10H2,1-6H3
SMILES CN(CCO[Si](C)(C)C(C)(C)C)c1ccc(O)cn1
IUPAC Name 6-[2-[tert-butyl(dimethyl)silyl]oxyethyl-methylamino]pyridin-3-ol
Molecular Weight (g/mol) 282.45
Formula Weight 282.1764