Learn More
6-[[2-(tert-Butyldimethylsilyloxy)ethyl]methylamino]pyridin-3-ol, TRC
Specifications
Specifications
| Chemical Name or Material | 6-[[2-(tert-Butyldimethylsilyloxy)ethyl]methylamino]pyridin- |
| CAS | 326496-02-0 |
| Molecular Formula | C14 H26 N2 O2 Si |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C14H26N2O2Si/c1-14(2,3)19(5,6)18-10-9-16(4)13-8-7-12(17)11-15-13/h7-8,11,17H,9-10H2,1-6H3 |
| SMILES | CN(CCO[Si](C)(C)C(C)(C)C)c1ccc(O)cn1 |
| IUPAC Name | 6-[2-[tert-butyl(dimethyl)silyl]oxyethyl-methylamino]pyridin-3-ol |
| Molecular Weight (g/mol) | 282.45 |
| Formula Weight | 282.1764 |