Learn More
(5S,5’S)-Dihydroxy Lysinonorleucine, TRC
Specifications
Specifications
| Chemical Name or Material | (5S,5'S)-Dihydroxy Lysinonorleucine |
| CAS | 32619-23-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C12 H25 N3 O6 |
| Quantity | 5 mg |
| InChI Formula | InChI=1S/C12H25N3O6/c13-9(11(18)19)3-1-7(16)5-15-6-8(17)2-4-10(14)12(20)21/h7-10,15-17H,1-6,13-14H2,(H,18,19)(H,20,21)/t7?,8?,9-,10-/m0/s1 |
| SMILES | N[C@@H](CCC(O)CNCC(O)CC[C@H](N)C(=O)O)C(=O)O |
| IUPAC Name | (2S)-2-amino-6-[[(5S)-5-amino-5-carboxy-2-hydroxypentyl]amino]-5-hydroxyhexanoic acid |
| Molecular Weight (g/mol) | 307.34 |
| Formula Weight | 307.1743 |
| Show More |