missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Chloro-4-morpholinophenyl)-N'-hydroxyformimidamide, MedChemExpress

Product Code. 16419237
Encompass_Preferred
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 16419237

Brand: Medchem Express HY15603

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TS-011 is a selective inhibitor of 20-Hydroxyeicosatetraenoic acid synthesis.

TS-011 is a selective inhibitor of 20-Hydroxyeicosatetraenoic acid synthesis.

Specifications

Chemical Name or Material N-(3-Chloro-4-morpholinophenyl)-N'-hydroxyformimidamide
Physical Form Solid
CAS 339071-18-0
Color Light Nude
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C11H14ClN3O2
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 250 mg/mL (977.71 mM; Need ultrasonic)
SMILES ClC(C=C(/N=C/NO)C=C1)=C1N2CCOCC2
Molecular Weight (g/mol) 255.7
Formula Weight 255.7
Percent Purity 99.08%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.