missing translation for 'onlineSavingsMsg'
Learn More

PF-2771, MedChemExpress

Product Code. 16430258
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16430258 5 mg 5mg
16497788 10 mg 10mg
16477626 100 mg 100mg
16463747 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16430258 Supplier Medchem Express Supplier No. HY195305mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-2771 is a potent and selective centromere protein E (CENP-E) inhibitor, inhibiting CENP-E motor activity with an IC50 of 16.1 nM; PF-2771 is used as an anticancer agent.

PF-2771 is a potent and selective centromere protein E (CENP-E) inhibitor, inhibiting CENP-E motor activity with an IC50 of 16.1 nM

Specifications

Chemical Name or Material PF-2771
CAS 2070009-55-9
Color White
Purity Grade Notes Research
Molecular Formula C29H36ClN5O4
Quantity 5 mg
Solubility Information DMSO : ≥ 32 mg/mL (57.75 mM)
SMILES ClC1=CC(C(N[C@@H](CC2=CC=C(C3=CN(C)C(C(C)=O)=N3)C=C2)CNC(CN(C)C)=O)=O)=CC=C1OC(C)C
Molecular Weight (g/mol) 554.08
Formula Weight 554.08
Percent Purity 96.44%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.