missing translation for 'onlineSavingsMsg'
Learn More

Helioxanthin derivative 5-4-2, MedChemExpress

Product Code. 16409667
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. Quantity unitSize
16409667 5 mg 5mg
16407128 10 mg 10mg
2 options
This item is not returnable. View return policy

Product Code. 16409667

Brand: Medchem Express HY16679

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Helioxanthin derivative 5-4-2 is an analogue of helioxanthin, exhibites significant in vitro anti-HBV activity with EC50 of 0.08 uM in HepG2.2.15 cells.

Helioxanthin derivative 5-4-2 is an analogue of helioxanthin, exhibites significant in vitro anti-HBV activity with EC50 of 0.08 uM in HepG2.2.15 cells.

Specifications

Chemical Name or Material Helioxanthin derivative 5-4-2
CAS 203935-39-1
Color White
Purity Grade Notes Research
Molecular Formula C20H13NO5
Quantity 5 mg
Synonym Helioxanthin 5-4-2
Solubility Information DMSO : 50 mg/mL (143.96 mM; Need ultrasonic)
SMILES O=C1NCC2=C(C3=CC=C(OCO4)C4=C3)C5=C6C(OCO6)=CC=C5C=C21
Molecular Weight (g/mol) 347.32
Formula Weight 347.32
Percent Purity 98.22%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.