missing translation for 'onlineSavingsMsg'
Learn More

Theophylline, Mouse, Clone: 11-84.4.1, Abnova™

Product Code. 16075486
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16075486 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16075486 Supplier Abnova Supplier No. MAB2230.100ug
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse monoclonal antibody raised against Theophylline.

Specifications

Antigen Theophylline
Applications ELISA, Radioimmune Assays (RIA)
Classification Monoclonal
Clone 11-84.4.1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Theophylline.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Host Species Mouse
Immunogen Theophylline conjugated with KLH.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity These antibodies recognize theophylline and cross-react to different degrees with caffeine: MAB2228 (∼2%)<MAB2229<MAB2230<MAB2231<MAB2232 (∼25%).
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.