Learn More
Novus Biologicals™ KIR2DL1/KIR2DS5 Antibody (Innate patent anti-KIR2DL) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28027-50ug
 View more versions of this product
Product Code. 30046987
  • 213.00 € / 50µg
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
KIR2DL1/KIR2DS5 Monoclonal antibody specifically detects KIR2DL1/KIR2DS5 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KIR2DL1/KIR2DS5
Monoclonal
Unconjugated
P43626
Human
Protein A purified
RUO
3802
Human
Purified
Flow Cytometry, ELISA, Functional Assay
Innate patent anti-KIR2DL
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
CD158a
KIR2DL1/ CD158a
50 μg
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1