Learn More
Novus Biologicals™ Cannabinoid R1/CB1/CNR1 Antibody (GFB-024) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28839-50ug
 View more versions of this product
Product Code. 30045313
  • 213.00 € / 50µg
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
Cannabinoid R1/CB1/CNR1 Monoclonal antibody specifically detects Cannabinoid R1/CB1/CNR1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cannabinoid R1/CB1/CNR1
Monoclonal
Unconjugated
Q96F85
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
GFB-024
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
CANN6CB1R, cannabinoid receptor 1 (brain), CB1A, CB1K5, CB-RCB1cannabinoid receptor 1, central cannabinoid receptor, CNR
CB1/ CNR1
50 μg
Cancer, GPCR, Neuroscience, Neurotransmission, Vision
1268
Human
Purified