Learn More
Agilent Technologies 50mm Bor. buffer, ph9.3+100mm SDS, 250ml
Brand: Agilent Technologies 5062-8574
50mm Bor. buffer, ph9.3+100mm SDS, 250ml
Safety and Handling
- 50mm Bor. buffer, ph9.3+100mm SDS
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360-May damage fertility or the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Sodium dodecyl sulphate, Disodium tetraborate, anhydrous
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.