missing translation for 'onlineSavingsMsg'
Learn More

SCH900776, MedChemExpress

Product Code. 16432697
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16432697

Brand: Medchem Express HY15532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SCH900776 (MK-8776) is a potent, selective and orally bioavailable inhibitor of checkpoint kinase1 (Chk1) with an IC50 of 3 nM. SCH900776 shows 50- and 500-fold selectivity over CDK2 and Chk2, respectively.

SCH900776 (MK-8776) is a potent, selective and orally bioavailable inhibitor of checkpoint kinase1 (Chk1) with an IC50 of 3 nM. SCH900776 shows 50- and 500-fold selectivity over CDK2 and Chk2, respectively.

Specifications

Chemical Name or Material SCH900776
Physical Form Solid
CAS 891494-63-6
Color Earth Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C15H18BrN7
For Use With (Application) Cancer-Kinase/protease
Synonym MK-8776
Solubility Information DMSO : ≥ 100 mg/mL (265.78 mM)
SMILES NC1=C(Br)C([C@H]2CNCCC2)=NC3=C(C4=CN(C)N=C4)C=NN13
Molecular Weight (g/mol) 376.25
Formula Weight 376.25
Percent Purity 98.71%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.