missing translation for 'onlineSavingsMsg'
Learn More

NCB-0846, MedChemExpress

Product Code. 16429736
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16429736 50 mg 50mg
16472656 25 mg 25mg
16444466 100 mg 100mg
16435917 5 mg 5mg
16413188 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16429736 Supplier Medchem Express Supplier No. HY100830

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NCB-0846 is an orally available TNIK inhibitor with an IC50 of 21 nM.

NCB-0846 is an orally available TNIK inhibitor with an IC50 of 21 nM.

Specifications

Chemical Name or Material NCB-0846
CAS 1792999-26-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H21N5O2
Quantity 50 mg
Solubility Information DMSO : ≥ 30 mg/mL (79.91 mM)
SMILES O[C@@H](CC1)CC[C@@H]1OC2=CC=CC(C=N3)=C2N=C3NC4=CC=C(N=CN5)C5=C4
Molecular Weight (g/mol) 375.42
Formula Weight 375.42
Percent Purity 98.22%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.