missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc, Zn/Al, Zamak 3, Z33520, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Zinc, Zn/Al, Zamak 3, Z33520, 50g
Specifications
Specifications
| Chemical Name or Material | Silicon/Cadmium/Lanthanum/Nickel/Lead/Cerium/Manganese/Iron/Magnesium/Copper/Aluminium/Tin |
| CAS | 7440-66-6 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Silicon 0.0011 %/Cadmium 0.0023 %/Lanthanum 0.0029 %/Nickel 0.003 %/Lead 0.0033 %/Cerium 0.0067 %/Manganese 0.0077 %/Iron 0.0144 %/Magnesium 0.0602 %/Copper 0.1 %/Aluminium 3.53 %/Tin <0.001 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction