Learn More
Pan Biotech 500ML MEM Eagle w: EBSS, w: stable Glutamine, w:2.2 g/L NaHCO3
Brand:  Pan Biotech P04-09500
Encompass_Preferred
500ML MEM Eagle w: EBSS, w: stable Glutamine, w:2.2 g/L NaHCO3
Product Code. 17190849
  • 18.51 € / 500mL