missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-Lansoprazole, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
(R)-Lansoprazole is the R enantiomer of Lansoprazole, Lansoprazole (AG 1749) is an orally active proton pump inhibitor which prevents the stomach from producing acid. Lansoprazole (AG 1749) is a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor (exosome inhibitor).
Specifications
Specifications
| Chemical Name or Material | (R)-Lansoprazole |
| CAS | 138530-94-6 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C16H14F3N3O2S |
| Quantity | 500 mg |
| Synonym | Dexlansoprazole |
| Solubility Information | DMSO : 100 mg/mL (270.74 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.27 mM; Need ultrasonic) |
| SMILES | O=[S@@](C1=NC2=CC=CC=C2N1)CC3=NC=CC(OCC(F)(F)F)=C3C |
| Molecular Weight (g/mol) | 369.36 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction