missing translation for 'onlineSavingsMsg'
Learn More

Fluconazole, MedChemExpress

Product Code. 16422016
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16422016

Brand: Medchem Express HYB0101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fluconazole (UK-49858) is a triazole antifungal agent with excellent activities against a broad range of fungi, especially against Candida albicans. Fluconazole inhibits C. albicans and Candida kefyr with IC99s range from 0.20 μg/mL to 0.39 μg/mL.

Fluconazole (UK-49858) is a triazole antifungal agent with excellent activities against a broad range of fungi, especially against Candida albicans. Fluconazole inhibits C. albicans and Candida kefyr with IC99s range from 0.20 μg/mL to 0.39 μg/mL.

Specifications

Chemical Name or Material Fluconazole
Physical Form Powder
CAS 86386-73-4
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C13H12F2N6O
For Use With (Application) COVID-19-immunoregulation
Synonym UK-49858
Solubility Information DMSO : ≥ 100 mg/mL (326.51 mM) ∣H2O : 2 mg/mL (6.53 mM; Need ultrasonic)
SMILES FC1=CC=C(C(O)(CN2C=NC=N2)CN3C=NC=N3)C(F)=C1
Molecular Weight (g/mol) 306.27
Formula Weight 306.27
Percent Purity 99.21%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.