missing translation for 'onlineSavingsMsg'
Learn More

Ferulic acid, MedChemExpress

Product Code. 16422346
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16422346

Brand: Medchem Express HYN0060

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Ferulic acid is a novel fibroblast growth factor receptor 1 (FGFR1) inhibitor with IC50s of 3.78 and 12.5 μM for FGFR1 and FGFR2, respectively.

Ferulic acid is a novel fibroblast growth factor receptor 1 (FGFR1) inhibitor with IC50s of 3.78 and 12.5 μM for FGFR1 and FGFR2, respectively.

Specifications

Chemical Name or Material Ferulic acid
CAS 1135-24-6
Color White
Purity Grade Notes Research
Molecular Formula C10H10O4
Quantity 500 mg
Synonym Coniferic acid
Solubility Information DMSO : 100 mg/mL (514.99 mM; Need ultrasonic)
SMILES O=C(O)/C=C/C1=CC=C(O)C(OC)=C1
Molecular Weight (g/mol) 194.18
Formula Weight 194.18
Percent Purity 98.57%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.