Learn More
Hamilton™ 50 μL 700 Series Microliter Syringe
Product Code. 17864212
Click to view available options
Needle Gauge:
22s G
Custom
No Needle Included
Needle Point Style:
3
Blunt
Custom
No Needle Included
Sharp, Bevel, Curved, Non-coring
Connection Type:
Cemented
Removable
Unit Size:
Each
Product Code. 17864212
Brand: Hamilton™ CAL765501
Please to purchase this item. Need a web account? Register with us today!
The 700 Series Microliter Syringe fulfi the general liquid handling requirements of manufacturing and research laboratories.
  • The 700 series is the original Hamilton syringe.
  • This series of Microliter syringes is designed to solve the general liquid handling requirements of manufacturing and research laboratories.
  • The 700 series remains the industry standard.
  • The plungers and barrels are not interchangeable or replaceable.
  • Dispenses volumes up to 50 μL.
Specifications
Autoclavable Autoclavable
Material Stainless Steel
Needle Gauge No Needle Included
Diameter (English) Inner 0.041 in.
Diameter (English) Outer 0.305 in.
Diameter (Metric) Inner 1.03 mm
Diameter (Metric) Outer 7.75 mm
Length (English) Needle No Needle Included
Product Type Microliter Syringe
Max. Temperature (English) 239°F
Max. Temperature (Metric) 115°C
Min. Temperature (English) 50°F
Model 705 RN
Min. Temperature (Metric) 10°C
Quantity 1
Volume (Metric) 50 μL
Length (Metric) Needle No Needle Included
Needle Point Style No Needle Included
Connection Type Removable