Learn More
5'-Tosyl-2'-deoxy Cytidine, TRC
Specifications
Specifications
| Chemical Name or Material | 5'-Tosyl-2'-deoxy Cytidine |
| CAS | 27999-55-9 |
| Molecular Formula | C16 H19 N3 O6 S |
| Quantity | 10 mg |
| Synonym | 5-Tosyl-2-Deoxy Cytidine |
| InChI Formula | InChI=1S/C16H19N3O6S/c1-10-2-4-11(5-3-10)26(22,23)24-9-13-12(20)8-15(25-13)19-7-6-14(17)18-16(19)21/h2-7,12-13,15,20H,8-9H2,1H3,(H2,17,18,21)/t12-,13+,15+/m0/s1 |
| SMILES | Cc1ccc(cc1)S(=O)(=O)OC[C@H]2O[C@H](C[C@@H]2O)N3C=CC(=NC3=O)N |
| IUPAC Name | [(2R,3S,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methyl 4-methylbenzenesulfonate |
| Molecular Weight (g/mol) | 381.4 |
| Formula Weight | 381.0995 |