Learn More
Alfa Aesar™ 5-tert-Butyl-2-methoxyaniline, 97%
Product Code. p-7036686
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15401138 5 g 5g
15411138 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15401138 Supplier Alfa Aesar™ Supplier No. H55019.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 3535-88-4
Molecular Formula C11H17NO
Molecular Weight (g/mol) 179.263
MDL Number MFCD00026299
InChI Key DLTLFDQLMFMTRQ-UHFFFAOYSA-N
Synonym 5-tert-butyl-2-methoxyaniline, benzenamine, 5-1,1-dimethylethyl-2-methoxy, 4-tert-butyl-o-anisidine, 2-amine-4-tert-butylanisole, 5-tert-butyl-o-anisidine, 5-tert-butyl-2-methoxy-phenylamine, 5-tert-butyl-2-methoxyphenylamine
PubChem CID 4176318
IUPAC Name 5-tert-butyl-2-methoxyaniline
SMILES CC(C)(C)C1=CC(=C(C=C1)OC)N
Specifications
Melting Point 29°C to 30°C
Density 0.988
Boiling Point 204°C to 208°C
Flash Point 113°C (235°F)
Quantity 5 g
Formula Weight 179.26
Percent Purity 97%
Chemical Name or Material 5-tert-Butyl-2-methoxyaniline