Learn More
5’-Sulfamoyladenosine, TRC
Product Code. 30411336
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30411336 10 mg
30419974 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30411336 Supplier Toronto Research Chemicals Supplier No. TRCS78900010MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25030-31-3
Molecular Formula C10H14N6O6S
Molecular Weight (g/mol) 346.32
Synonym 5'-Ester with Adenosine Sulfamic Acid, 5'-O-Sulfamoyladenosine, NSC 133114
IUPAC Name ((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl sulfamate
SMILES NC1=NC=NC2=C1N=CN2[C@H]3[C@H](O)[C@H](O)[C@@H](COS(N)(=O)=O)O3
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C10H14N6O6S/c11-8-5-9(14-2-13-8)16(3-15-5)10-7(18)6(17)4(22-10)1-21-23(12,19)20/h2-4,6-7,10,17-18H,1H2,(H2,11,13,14)(H2,12,19,20)/t4-,6-,7-,10-/m1/s1
Formula Weight 346.07
Percent Purity >95%
Chemical Name or Material 5'-Sulfamoyladenosine
Grade HPLC