Learn More
Invitrogen™ 5-ROX (5-Carboxy-X-Rhodamine, Triethylammonium Salt), single isomer
Description
The carboxylic acid of 5-ROX is used for oligonucleotide labeling and automated DNA sequencing applications. Conjugates of this dye have longer-wavelength spectra than the spectra of Lisaamine™ rhodamine B conjugates, but somewhat shorter-wavelength spectra than those of Texas Red™ conjugates.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | 5-ROX |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Chemical Reactivity | Amine |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Carboxylic Acid |
| Label or Dye | ROX |
| Label Type | Classic Dyes |