Learn More
5'-O-(p-Toluenesulfonyl)-2'-deoxyuridine, TRC
Specifications
Specifications
| Chemical Name or Material | 5'-O-(p-Toluenesulfonyl)-2'-deoxyuridine |
| CAS | 27999-47-9 |
| Molecular Formula | C16H18N2O7S |
| Quantity | 100 mg |
| Synonym | 1-(5-O-Tosyl-2-deoxy-beta-D-erythro-pentofuranosyl)uracil, 2'-Deoxy-5'-O-p-toluenesulfonyluridine, 5'-O-Tosyl-2'-deoxyuridine, 5'-O-p-Tolylsulfonyl-2'-deoxyuridine |
| InChI Formula | InChI=1S/C16H18N2O7S/c1-10-2-4-11(5-3-10)26(22,23)24-9-13-12(19)8-15(25-13)18-7-6-14(20)17-16(18)21/h2-7,12-13,15,19H,8-9H2,1H3,(H,17,20,21)/t12-,13+,15+/m0/s1 |
| SMILES | Cc1ccc(cc1)S(=O)(=O)OC[C@H]2O[C@H](C[C@@H]2O)N3C=CC(=O)NC3=O |
| IUPAC Name | [(2R,3S,5R)-5-(2,4-dioxopyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methyl 4-methylbenzenesulfonate |
| Molecular Weight (g/mol) | 382.39 |
| Formula Weight | 382.0835 |