Learn More
(5-Nitro-1-naphthyl)amine, TRC
Specifications
Specifications
| Chemical Name or Material | (5-Nitro-1-naphthyl)amine |
| CAS | 3272-91-1 |
| Molecular Formula | C10H8N2O2 |
| Quantity | 2.5 g |
| InChI Formula | InChI=1S/C10H8N2O2/c11-9-5-1-4-8-7(9)3-2-6-10(8)12(13)14/h1-6H,11H2 |
| SMILES | C1=CC2=C(C=CC=C2[N+](=O)[O-])C(=C1)N |
| IUPAC Name | 5-nitronaphthalen-1-amine |
| Molecular Weight (g/mol) | 188.18 |
| Formula Weight | 188.0586 |