Learn More
5-(Morpholin-4-ylmethyl)quinolin-8-ol, TRC
Specifications
Specifications
| Chemical Name or Material | 5-(Morpholin-4-ylmethyl)quinolin-8-ol |
| CAS | 312611-90-8 |
| Molecular Formula | C14 H16 N2 O2 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C14H16N2O2/c17-13-4-3-11(10-16-6-8-18-9-7-16)12-2-1-5-15-14(12)13/h1-5,17H,6-10H2 |
| SMILES | Oc1ccc(CN2CCOCC2)c3cccnc13 |
| IUPAC Name | 5-(morpholin-4-ylmethyl)quinolin-8-ol |
| Molecular Weight (g/mol) | 244.289 |
| Formula Weight | 244.121 |