missing translation for 'onlineSavingsMsg'
Learn More

5-Methylnicotinic acid, 97%, Thermo Scientific™

Catalog Number p-7077427
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
3222-49-9
C7H7NO2
137.14
DJDHHXDFKSLEQY-UHFFFAOYSA-N
5-methylnicotinic acid, 5-methyl-nicotinic acid, 5-methyl nicotinic acid, 5-methylnicotic acid, 5-methyl-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 5-methyl, 5-methyinicotinic acid, pubchem11961, 5-methyl nicotinic aicd, acmc-1afvp
256208
5-methylpyridine-3-carboxylic acid
CC1=CN=CC(=C1)C(=O)O
This item is not returnable. View return policy
3222-49-9
C7H7NO2
137.14
DJDHHXDFKSLEQY-UHFFFAOYSA-N
5-methylnicotinic acid, 5-methyl-nicotinic acid, 5-methyl nicotinic acid, 5-methylnicotic acid, 5-methyl-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 5-methyl, 5-methyinicotinic acid, pubchem11961, 5-methyl nicotinic aicd, acmc-1afvp
256208
5-methylpyridine-3-carboxylic acid
CC1=CN=CC(=C1)C(=O)O
Melting Point 211.0°C to 213.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 137.14
Percent Purity 97%
Chemical Name or Material 5-Methylnicotinic acid
compliance-icons
Product Identifier
  • 5-Methylnicotinic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'