Learn More
5-Methyl-3,4-diphenylisoxazole, TRC
Product Code. 30464723
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30464723 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30464723 Supplier Toronto Research Chemicals Supplier No. TRCM312540500MG
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
CAS 37928-17-9
Molecular Formula C16 H13 N O
Quantity 500 mg
Synonym Parecoxib TLC impurity 19, Desulfonamide valdecoxib TLC impurity 10
InChI Formula InChI=1S/C16H13NO/c1-12-15(13-8-4-2-5-9-13)16(17-18-12)14-10-6-3-7-11-14/h2-11H,1H3
SMILES Cc1onc(c2ccccc2)c1c3ccccc3
IUPAC Name 5-methyl-3,4-diphenyl-1,2-oxazole
Molecular Weight (g/mol) 235.28
Formula Weight 235.0997
Chemical Name or Material 5-Methyl-3,4-diphenylisoxazole