missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™

Product Code. 11411370
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11411370 1 g 1g
11421370 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11411370

Brand: Thermo Scientific Alfa Aesar H26411.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Methyl-2-(trifluoromethyl)benzaldehyde

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886502-76-7
Molecular Formula C9H7F3O
Molecular Weight (g/mol) 188.149
MDL Number MFCD06660294
InChI Key ZXQHVZWHSXZEQV-UHFFFAOYSA-N
Synonym 5-methyl-2-trifluoromethyl benzaldehyde, 5-methyl-2-trifluormethyl benzaldehyde, 5-methyl-2-trifluoromethyl-benzaldehyde
PubChem CID 17750727
IUPAC Name 5-methyl-2-(trifluoromethyl)benzaldehyde
SMILES CC1=CC(=C(C=C1)C(F)(F)F)C=O

Specifications

Refractive Index 1.471
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 188.15
Percent Purity 97%
Chemical Name or Material 5-Methyl-2-(trifluoromethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.