Learn More
5-Methyl-1H-pyrazole Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Methyl-1H-pyrazole Hydrochloride |
| CAS | 119760-62-2 |
| Molecular Formula | C4 H6 N2 . H Cl |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C4H6N2.ClH/c1-4-2-3-5-6-4;/h2-3H,1H3,(H,5,6);1H |
| SMILES | Cl.Cc1ccn[nH]1 |
| IUPAC Name | 5-methyl-1H-pyrazole;hydrochloride |
| Molecular Weight (g/mol) | 118.565 |
| Formula Weight | 118.03 |