Learn More
5-Isopropyl-3-methylphenol, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Isopropyl-3-methylphenol |
| CAS | 3228-03-3 |
| Molecular Formula | C10H14O |
| Quantity | 2.5 g |
| Synonym | 3-Methyl-5-(1-methylethyl)phenol, 5-isopropyl-m-Cresol, m-Cymen-5-ol, 3-Isopropyl-5-methylphenol, 3-Methyl-5-(propan-2-yl)phenol, 3-Methyl-5-isopropylphenol, 5-Isopropyl-3-methylphenol, 5-Isopropyl-m-cresol |
| InChI Formula | InChI=1S/C10H14O/c1-7(2)9-4-8(3)5-10(11)6-9/h4-7,11H,1-3H3 |
| SMILES | OC=1C=C(C=C(C1)C(C)C)C |
| IUPAC Name | 3-methyl-5-propan-2-ylphenol |
| Molecular Weight (g/mol) | 150.22 |
| Formula Weight | 150.1045 |