missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy Tryptophol Beta-D-Glucuronide Sodium Salt, TRC

Product Code. 30399045
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
10 mg
Unit Size:
0.5mg
10mg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399045 10 mg 10mg
30394000 1 mg 1mg
30400511 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30399045 Supplier Toronto Research Chemicals Supplier No. TRCH97601010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Hydroxy Tryptophol b-D-Glucuronide Sodium Salt
Synonym 3-(2-Hydroxyethyl)-1H-indol-5-yl β-D-Glucopyranosiduronic Acid Sodium Salt ∣3-(2-Hydroxyethyl)indol-5-yl β-D-Glucosiduronic Acid Sodium Salt ∣
Quantity 10 mg
Shipping Condition Room Temperature
Molecular Formula C16H19NNaO8
Formula Weight 375.09
Purity >95% (HPLC)
Molecular Weight (g/mol) 375.31
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.