Learn More
5-Hydroxy Thiabendazole, TRC
Product Code. 30135101
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135101 25 mg 25mg
17960915 1 mg 1mg
17991385 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135101 Supplier Toronto Research Chemicals Supplier No. TRCH96120025MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 948-71-0
Molecular Formula C10 H7 N3 O S
Molecular Weight (g/mol) 217.25
Synonym 1H-Benzimidazol-5-ol, 2-(4-thiazolyl)- (9CI), 5(or 6)-Benzimidazolol, 2-(4-thiazolyl)- (7CI), 5-Benzimidazolol, 2-(4-thiazolyl)- (8CI), 2-(4-Thiazolyl)-1H-benzimidazol-6-ol, 5-Hydroxythiabendazole, Thiabendazole-5-hydroxy
IUPAC Name 2-(1,3-thiazol-4-yl)-1H-benzimidazol-5-ol
SMILES Oc1ccc2[nH]c(nc2c1)c3cscn3
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C10H7N3OS/c14-6-1-2-7-8(3-6)13-10(12-7)9-4-15-5-11-9/h1-5,14H,(H,12,13)
Formula Weight 217.031
Percent Purity >95%
Chemical Name or Material Thiabendazole-5-hydroxy
Grade HPLC