Learn More
5-Hydroxy Propranolol-d5, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Hydroxy Propranolol-d5 |
| CAS | 1185084-01-8 |
| Molecular Formula | C16H16D5NO3 |
| Quantity | 10 mg |
| Synonym | 5-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]-1-naphthalenol-d5, 5'-Hydroxypropranolol-d5 |
| InChI Formula | InChI=1S/C16H21NO3/c1-11(2)17-9-12(18)10-20-16-8-4-5-13-14(16)6-3-7-15(13)19/h3-8,11-12,17-19H,9-10H2,1-2H3/i9D2,10D2,12D |
| SMILES | CC(C)NC([2H])([2H])C(O)([2H])C([2H])([2H])OC1=CC=CC2=C1C=CC=C2O |
| Molecular Weight (g/mol) | 280.37 |
| Formula Weight | 280.1835 |