Learn More
5-Hydroxy Bromantane, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Hydroxy Bromantane |
| CAS | 560070-28-2 |
| Molecular Formula | C16H20BrNO |
| Quantity | 5 mg |
| Synonym | Tricyclo[3.3.1.13,7]decan-1-ol, 4-[(4-bromophenyl)amino]-, stereoisomer, Stereoisomer of 4-[(4-bromophenyl)amino]tricyclo[3.3.1.13,7]decan-1-ol, 5-Hydroxybromantan |
| InChI Formula | InChI=1S/C16H20BrNO/c17-13-1-3-14(4-2-13)18-15-11-5-10-6-12(15)9-16(19,7-10)8-11/h1-4,10-12,15,18-19H,5-9H2/t10-,11-,12+,15?,16- |
| SMILES | O[C@]12C[C@@H]3C[C@H](C1)C(Nc4ccc(Br)cc4)[C@@H](C3)C2 |
| IUPAC Name | (3S,5R)-4-(4-bromoanilino)adamantan-1-ol |
| Molecular Weight (g/mol) | 322.24 |
| Formula Weight | 321.0728 |