missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-HT1A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-03099
This item is not returnable.
View return policy
Description
5-HT1A Polyclonal specifically detects 5-HT1A in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| 5-HT1A | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| 5-HT-1A, 5-HT1a receptor, 5-HT1A5-hydroxytryptamine receptor 1A, 5-hydroxytryptamine (serotonin) receptor 1A, ADRB2RL15HT1a, ADRBRL1, G protein coupled receptor, G-21, guanine nucleotide-binding regulatory protein-coupled receptor, Serotonin receptor 1A | |
| A synthetic peptide of human 5-HT1A. | |
| 100 μL | |
| Cell Cycle and Replication, GPCR | |
| 3350 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction