Learn More
5-Formylfuran-2-boronic Acid Pinacol Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Formylfuran-2-boronic Acid Pinacol Ester |
| CAS | 273731-82-1 |
| Molecular Formula | C11H15BO4 |
| Quantity | 100 mg |
| Synonym | 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-furancarboxaldehyde |
| InChI Formula | InChI=1S/C11H15BO4/c1-10(2)11(3,4)16-12(15-10)9-6-5-8(7-13)14-9/h5-7H,1-4H3 |
| SMILES | CC1(C)OB(OC1(C)C)c2oc(C=O)cc2 |
| IUPAC Name | 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)furan-2-carbaldehyde |
| Molecular Weight (g/mol) | 222.05 |
| Formula Weight | 222.1063 |