Learn More
5-Fluoro-1H-indole-2,3-dione 3-Hydrazone, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Fluoro-1H-indole-2,3-dione 3-Hydrazone |
| CAS | 283584-52-1 |
| Molecular Formula | C8 H6 F N3 O |
| Quantity | 5 g |
| Synonym | Sunitinib Impurity 21 |
| InChI Formula | InChI=1S/C8H6FN3O/c9-4-1-2-6-5(3-4)7(12-10)8(13)11-6/h1-3H,10H2,(H,11,12,13) |
| SMILES | N\N=C\1/C(=O)Nc2ccc(F)cc12 |
| IUPAC Name | (3Z)-5-fluoro-3-hydrazinylidene-1H-indol-2-one |
| Molecular Weight (g/mol) | 179.151 |
| Formula Weight | 179.049 |