Learn More
5-Ethoxycarbonyl SU 5402 Methyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 2-[(1,2-Dihydro-2-oxo-3H-indol-3-ylidene)methyl]-5-(ethoxycarbonyl)-4-methyl-1H-pyrrole-3-propanoic Acid Methyl Ester |
| CAS | 258831-76-4 |
| Molecular Formula | C21H22N2O5 |
| Quantity | 25 mg |
| Synonym | 5-Ethoxycarbonyl SU 5402 Methyl Ester, 2-[(1,2-Dihydro-2-oxo-3H-indol-3-ylidene)methyl]-5-(ethoxycarbonyl)-4-methyl-1H-pyrrole-3-propanoic Acid Methyl Ester |
| InChI Formula | InChI=1S/C21H22N2O5/c1-4-28-21(26)19-12(2)13(9-10-18(24)27-3)17(22-19)11-15-14-7-5-6-8-16(14)23-20(15)25/h5-8,11,22H,4,9-10H2,1-3H3,(H,23,25)/b15-11- |
| SMILES | CCOC(=O)c1[nH]c(\C=C\2/C(=O)Nc3ccccc23)c(CCC(=O)OC)c1C |
| IUPAC Name | ethyl 4-(3-methoxy-3-oxopropyl)-3-methyl-5-[(Z)-(2-oxo-1H-indol-3-ylidene)methyl]-1H-pyrrole-2-carboxylate |
| Molecular Weight (g/mol) | 382.41 |
| Formula Weight | 382.1529 |