Learn More
5-Desthiopropyl-5-hydroxy-ticagrelor, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Desthiopropyl-5-hydroxy-ticagrelor |
| CAS | 1644461-81-3 |
| Molecular Formula | C20 H22 F2 N6 O5 |
| Quantity | 1 mg |
| Synonym | Ticagrelor Related Compound 15, (1S,2S,3R,5S)-3-(7-(((1R,2S)-2-(3,4-Difluorophenyl)cyclopropyl)amino)-5-hydroxy-3H-[1,2,3]triazolo[4,5-d]pyrimidin-3-yl)-5-(2-hydroxyethoxy)cyclopentane-1,2-diol |
| InChI Formula | InChI=1S/C20H22F2N6O5/c21-10-2-1-8(5-11(10)22)9-6-12(9)23-18-15-19(25-20(32)24-18)28(27-26-15)13-7-14(33-4-3-29)17(31)16(13)30/h1-2,5,9,12-14,16-17,29-31H,3-4,6-7H2,(H2,23,24,25,32)/t9-,12+,13+,14-,16-,17+/m0/s1 |
| SMILES | OCCO[C@H]1C[C@H]([C@H](O)[C@@H]1O)n2nnc3c(N[C@@H]4C[C@H]4c5ccc(F)c(F)c5)nc(O)nc23 |
| IUPAC Name | (1S,2S,3R,5S)-3-[7-[[(1R,2S)-2-(3,4-difluorophenyl)cyclopropyl]amino]-5-hydroxy-triazolo[4,5-d]pyrimidin-3-yl]-5-(2-hydroxyethoxy)cyclopentane-1,2-diol |
| Molecular Weight (g/mol) | 464.42 |
| Formula Weight | 464.162 |