missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorooxindole, 98%

Product Code. 11469486
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469486 100 g 100g
11449486 5 g 5g
11459486 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469486 Supplier Thermo Scientific Alfa Aesar Supplier No. A19210.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 17630-75-0 | C8H6ClNO | 167.59 g/mol

5-Chloro-2-oxindole was used for the quantitation of 5-chloro-2-oxindole, concomitantly with all of its potential positional isomers using a single, highly specific, normal-phase chromatographic system. 5-Chloro-2-oxindole (5-Chlorooxindole) is a starting material for tenidap sodium, a pharmaceutical drug candidate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-2-oxindole was used for the quantitation of 5-chloro-2-oxindole, concomitantly with all of its potential positional isomers using a single, highly specific, normal-phase chromatographic system. 5-Chloro-2-oxindole (5-Chlorooxindole) is a starting material for tenidap sodium, a pharmaceutical drug candidate.

Solubility
Slightly Soluble (2.0 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17630-75-0
Molecular Formula C8H6ClNO
Molecular Weight (g/mol) 167.59
MDL Number MFCD00191927
InChI Key WWJLCYHYLZZXBE-UHFFFAOYSA-N
Synonym 5-chlorooxindole, 5-chloroindolin-2-one, 5-chloro-1,3-dihydro-2h-indol-2-one, 5-chloro-2-indolinone, 5-chloro-1,3-dihydro-indol-2-one, 5-chloro-2-oxindole, 2h-indol-2-one, 5-chloro-1,3-dihydro, 5-chloro-2,3-dihydro-1h-indol-2-one, 2-indolinone, 5-chloro, 5-chlorooxindol
PubChem CID 152801
SMILES ClC1=CC=C2NC(=O)CC2=C1

Specifications

Melting Point 196°C to 200°C
Quantity 100 g
IUPAC Name 5-chloro-1,3-dihydroindol-2-one
Formula Weight 167.6
Percent Purity 98%
Chemical Name or Material 5-Chlorooxindole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.