Learn More
5-Chloronaphthalen-1-amine, TRC
Specifications
Specifications
| CAS | 2750-80-3 |
| Molecular Formula | C10H8ClN |
| Quantity | 10 mg |
| Synonym | 5-Chloro-1-naphthalenamine, 5-Chloro-1-naphthylamine |
| InChI Formula | InChI=1S/C10H8ClN/c11-9-5-1-4-8-7(9)3-2-6-10(8)12/h1-6H,12H2 |
| SMILES | Nc1cccc2c(Cl)cccc12 |
| IUPAC Name | 5-chloronaphthalen-1-amine |
| Molecular Weight (g/mol) | 177.63 |
| Formula Weight | 177.0345 |
| Chemical Name or Material | 5-Chloronaphthalen-1-amine |